8.14528
8-Hydroxyquinoline sulfate monohydrate
for synthesis
Synonym(s):
8-Hydroxyquinoline sulfate monohydrate, Quinosol
About This Item
Recommended Products
Quality Level
form
powder
potency
1200 mg/kg LD50, oral (Rat)
>4000 mg/kg LD50, skin (Rat)
pH
3.2 (20 °C in H2O)
mp
182 °C
storage temp.
2-30°C
SMILES string
[S](=O)(=O)(O)O.n3c4c(ccc3)cccc4O.n1c2c(ccc1)cccc2O.O
InChI
1S/2C9H7NO.H2O4S.H2O/c2*11-8-5-1-3-7-4-2-6-10-9(7)8;1-5(2,3)4;/h2*1-6,11H;(H2,1,2,3,4);1H2
InChI key
BNCXJZDIJIVJJO-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service