Skip to Content
Merck

8.09691

m-Cresol

for synthesis

Synonym(s):

m-Cresol, 3-Hydroxytoluol, 3-Methylphenol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
3-(CH3)C6H4OH
CAS Number:
Molecular Weight:
108.14
UNSPSC Code:
12352104
EC Index Number:
203-577-9
NACRES:
NA.22
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

0.065 hPa ( 20 °C)

Quality Level

assay

≥99.0% (GC)

form

liquid

autoignition temp.

626 °C

potency

242 mg/kg LD50, oral (Rat)

expl. lim.

1.06 % (v/v)

pH

5 (20 °C, 20 g/L in H2O)

bp

203 °C/1013 hPa

mp

11.5 °C

transition temp

flash point 86 °C

solubility

23.5 g/L

density

1.03 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

Oc1cc(ccc1)C

InChI

1S/C7H8O/c1-6-3-2-4-7(8)5-6/h2-5,8H,1H3

InChI key

RLSSMJSEOOYNOY-UHFFFAOYSA-N

Application

m-Cresol can be used as a starting material to synthesize:
  • Thymol via isopropylation reaction with 2-propanol in the presence of an acid catalyst.
  • Terpolymeric resin by an acid-catalyzed polycondensation reaction with hexamine and formaldehyde.
  • 4,7-Dimethylcoumarin via modified Pechmann condensation reaction with ethyl acetoacetate.

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.031 - 1.037
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.
Appearance: colorless to dark yellow/light brown. Material may darken with time.


Still not finding the right product?

Explore all of our products under m-Cresol


pictograms

Skull and crossbonesCorrosion

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 2

flash_point_f

186.8 °F - closed cup

flash_point_c

86 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Comparison of mesoporous and microporous solid acid catalysts for highly selective synthesis of thymol by vapor phase isopropylation of m-cresol
Selvaraj M and Kawi S
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 109(1-3), 458-469 (2008)
Synthesis, Chacterization, and Thermal Study of Terpolymeric Resin Derived from m-cresol, Hexamine and Formaldehyde
Khedkar KM, et al.
E-Journal of Chemistry, 9(4), 1911-1918 (2012)
Ganapati D Yadav et al.
The journal of physical chemistry. A, 109(48), 11080-11088 (2005-12-08)
The alkylation of m-cresol with isopropyl alcohol in the presence of novel superacidic catalysts named as UDCaT-4, UDCaT-5, and UDCaT-6 was investigated. The catalysts are modified versions of zirconia showing high catalytic activity, stability, and reusability in the presence of



Global Trade Item Number

SKUGTIN
809691000004022536402123
809691400004027269274504
809691903004022536668604
809691050004022536402147
809691010004022536402130
809691250004022536402161
809691999904022536402185
809691100004022536402154