Skip to Content
Merck

8.07582

Heptanoic acid

for synthesis

Synonym(s):

Heptanoic acid, Enanthic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3(CH2)5COOH
CAS Number:
Molecular Weight:
130.18
UNSPSC Code:
12352106
EC Index Number:
203-838-7
NACRES:
NA.22
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

1.35 hPa ( 20 °C)

Quality Segment

assay

≥99.0% (GC)

form

liquid

autoignition temp.

380 °C

potency

7000 mg/kg LD50, oral (Rat)

kinematic viscosity

5.00 cSt(20 °C)

mp

-8 °C

transition temp

flash point 117 °C

density

0.92 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[O-]C(=O)CCCCCC.[H+]

InChI

1S/C7H14O2/c1-2-3-4-5-6-7(8)9/h2-6H2,1H3,(H,8,9)

InChI key

MNWFXJYAOYHMED-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.917 - 0.919
Identity (IR): passes test


Still not finding the right product?

Explore all of our products under Heptanoic acid


pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

target_organs

Respiratory system

Storage Class

8A - Combustible, corrosive hazardous materials

wgk

WGK 1

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library