Skip to Content
Merck

Skip To

8.07488

Polyethylene glycol

greener alternative

Greener Alternative

1000, for synthesis

Synonym(s):

Poly(ethylene glycol), Polyethylene glycol 1000, Polyglycol, Polyethylene oxide, Polyoxy ethylene, PEG 1000, PEG

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
NACRES:
NA.22
EC Index Number:
500-038-2
UNSPSC Code:
13111021
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

<0.1 hPa ( 20 °C)

Quality Segment

form

liquid

autoignition temp.

420 °C

potency

28000 mg/kg LD50, oral (Rat), >20000 mg/kg LD50, skin (Rabbit)

greener alternative product characteristics

Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

pH

4-7 (20 °C, 100 g/L in H2O)

mp

33-40 °C

transition temp

flash point 260 °C

density

1.2 g/cm3 at 20 °C

greener alternative category

storage temp.

2-30°C

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. Polyethylene glycol (PEG) is an eco-friendly, biodegradable polymer widely used in pharmaceuticals and cosmetics. Its non-toxic nature and versatility make it a sustainable choice, derived from renewable resources, contributing to greener product formulations. Click here for more information.

Application

Polyethylene glycol 1000 for synthesis. CAS 25322-68-3, pH 4 - 7 (100 g/l, H₂O, 20 °C).

Analysis Note

Melting range (lower value): ≥ 33 °C
Melting range (upper value): ≤ 40 °C
Hydroxyl value: 107 - 118
Average molecular mass: 950 - 1050
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.190158.074868.07491
description

1000, for synthesis

description

3000, for synthesis

description

600, for synthesis

description

6000, for synthesis

form

liquid

form

liquid

form

liquid

form

solid

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

no temp limit

storage temp.

2-30°C

density

1.2 g/cm3 at 20 °C

density

1.21 g/cm3 at 20 °C

density

1.13 g/cm3 at 20 °C

density

1.2 g/cm3 at 20 °C

mp

33-40 °C

mp

55-58 °C

mp

17-22 °C

mp

58-63 °C

autoignition temp.

420 °C

autoignition temp.

420 °C

autoignition temp.

>320 °C (DIN 51794)

autoignition temp.

420 °C


Still not finding the right product?

Explore all of our products under Polyethylene glycol


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

281.5 °F - closed cup

flash_point_c

138.6 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters