Skip to Content
Merck

8.04604

Hydrazin hydrate solution

80% solution in water, for synthesis

Synonym(s):

Hydrazine hydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352402
NACRES:
NA.22
Form:
solution
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Hydrazin hydrate (80% solution in water), for synthesis

vapor pressure

13 hPa ( 20 °C)

Quality Segment

form

solution

autoignition temp.

310 °C

expl. lim.

4.7-99 % (v/v) refers to pure substance()

reaction suitability

reagent type: reductant

pH

10.6-10.7 (20 °C, 10 g/L in H2O)

bp

117-119 °C/1013 hPa

mp

-60 °C

density

1.02 g/cm3 at 20 °C

storage temp.

15-25°C

Application

Hydrazin hydrate (80% solution in water) can be used as a reducing agent for the reduction of exfoliated graphene oxide (GO), which is used for the preparation of GO nanocomposites. It is also used as a reagent in the synthesis of 3-amino-2-thiohydantoins by reacting with alkyl isothiocynatocarboxylates.


signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

wgk

WGK 3

flash_point_f

195.8 °F - closed cup - DIN 51758

flash_point_c

91 °C - closed cup - DIN 51758

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Synthetical Application of Alkyl 2-isothiocyanatocarboxylates. A Simple Synthesis of 5-Substituted-3-amino-2-thioxo-4-imidazolidinones (3-Amino-2-thiohydantoins)
Floch L, et al.
Molecules (Basel), 4(10), 279-286 (1999)
Hydroxypropyl-β-cyclodextrin-polyurethane/graphene oxide magnetic nanoconjugates as effective adsorbent for chromium and lead ions
Nasiri, Shohreh and Alizadeh, Nina
Carbohydrate Polymers, 259, 117731-117731 (2021)



Global Trade Item Number

SKUGTIN
804604000004022536385594
804604100004022536385617
804604025004022536385600
804604999904022536385631
804604901004022536546452
804604905504022536385624