Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
2-(OH)C6H4COCH3
CAS Number:
Molecular Weight:
136.15
UNSPSC Code:
12352115
EC Index Number:
204-288-0
NACRES:
NA.22
MDL number:
Assay:
≥95.0% (GC)
Form:
liquid
vapor pressure
0.27 hPa ( 20 °C)
Quality Level
assay
≥95.0% (GC)
form
liquid
potency
2127 mg/kg LD50, oral (Rat), >2000 mg/kg LD50, skin (Rat)
expl. lim.
0.98-11.8 % (v/v)
bp
212-214 °C/1013 hPa
mp
4-6 °C
transition temp
flash point 106 °C
solubility
0.2 g/L
density
1.13 g/cm3 at 20 °C
storage temp.
2-30°C
SMILES string
Oc1c(cccc1)C(=O)C
InChI
1S/C8H8O2/c1-6(9)7-4-2-3-5-8(7)10/h2-5,10H,1H3
InChI key
JECYUBVRTQDVAT-UHFFFAOYSA-N
Application
2′-Hydroxyacetophenone for synthesis. CAS 118-93-4, chemical formula 2-(OH)C₆H₄COCH₃.
Analysis Note
Assay (GC, area%): ≥ 95.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.130 - 1.133
Identity (IR): passes test
Density (d 20 °C/ 4 °C): 1.130 - 1.133
Identity (IR): passes test
Still not finding the right product?
Explore all of our products under 2′-Hydroxyacetophenone
Storage Class
10 - Combustible liquids
wgk
WGK 2
flash_point_f
222.8 °F
flash_point_c
106 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 8043100250 | 04022536384566 |
| 8043100000 | 04022536384542 |