Skip to Content
Merck

8.03559

Dibenzosuberone

for synthesis

Synonym(s):

Dibenzosuberone, 10,11-Dihydro-5H-dibenzo[a,d]cyclohepten-5-one

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C15H12O
CAS Number:
Molecular Weight:
208.26
UNSPSC Code:
12352115
EC Index Number:
214-912-3
NACRES:
NA.22
MDL number:
Assay:
≥97.0% (GC)
Form:
crystals
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97.0% (GC)

form

crystals

potency

2511 mg/kg LD50, oral (Rat), >2000 mg/kg LD50, skin (Rat)

bp

191-194 °C/7 hPa

mp

31-34 °C

transition temp

flash point 200 °C

solubility

0.03 g/L

density

1.16 g/cm3 at 25 °C

storage temp.

2-30°C

SMILES string

O=C1c2c(cccc2)CCc3c1cccc3

InChI

1S/C15H12O/c16-15-13-7-3-1-5-11(13)9-10-12-6-2-4-8-14(12)15/h1-8H,9-10H2

InChI key

BMVWCPGVLSILMU-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Melting range (lower value): ≥ 31 °C
Melting range (upper value): ≤ 34 °C
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.


Still not finding the right product?

Explore all of our products under Dibenzosuberone


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library