Skip to Content

8.03129

Diethylene glycol monobutyl ether

for synthesis

Synonym(s):

Diethylene glycol monobutyl ether, Butyl carbitol, 2-(2-Butoxyethoxy)ethanol, Butyl diglycol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
HOCH2CH2OCH2CH2O(CH2)3CH3
CAS Number:
Molecular Weight:
162.23
UNSPSC Code:
12352104
EC Index Number:
203-961-6
NACRES:
NA.22
MDL number:
Assay:
≥98% (GC)
Grade:
synthesis grade
Bp:
226-234 °C/1013 hPa
Vapor pressure:
<1 hPa ( 20 °C)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

synthesis grade

Quality Segment

vapor pressure

<1 hPa ( 20 °C)

assay

≥98% (GC)

form

liquid

autoignition temp.

210 °C

potency

5660 mg/kg LD50, oral (Rat), 4120 mg/kg LD50, skin (Rabbit)

expl. lim.

0.7-5.9 % (v/v)

dilution

(for synthesis)

pH

7 (20 °C)

kinematic viscosity

6.1 cSt(20 °C)

bp

226-234 °C/1013 hPa

mp

-68 °C

transition temp

flash point 105 °C

density

0.95 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(CCOCCCC)CCO

InChI

1S/C8H18O3/c1-2-3-5-10-7-8-11-6-4-9/h9H,2-8H2,1H3

InChI key

OAYXUHPQHDHDDZ-UHFFFAOYSA-N

Application


  • Biopolymeric Anticorrosion Coatings from Cellulose Nanofibrils and Colloidal Lignin Particles.: This study describes the development of a novel anticorrosion coating made from biopolymers, including a discussion on the use of diethylene glycol monobutyl ether as a solvent to disperse components uniformly (Dastpak et al., 2021).

  • The investigation of structural and thermosensitive properties of new phosphazene derivatives bearing glycol and amino acid.: This research article explores the properties of phosphazene derivatives that incorporate glycol structures, detailing methods like NMR and IR spectroscopy where diethylene glycol monobutyl ether may be employed as a solvent in the synthesis process (Uslu and Güvenaltın, 2010).

  • Diethylene glycol mono butyl ether concentrations in room air from application of cleaner formulations to hard surfaces.: This study measures the air concentrations of diethylene glycol monobutyl ether in indoor environments following its application in cleaning agents, demonstrating its volatility and potential inhalation exposure during use (Gibson et al., 1991).

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.951 - 0.954
Water (K. F.): ≤ 0.20 %
Peroxide (as H₂O₂): ≤ 0.05 %
Identity (IR): passes test


Still not finding the right product?

Explore all of our products under Diethylene glycol monobutyl ether


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

210.2 °F - closed cup

flash_point_c

99 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library