Skip to Content
Merck
All Photos(1)

Key Documents

8.02254

Sigma-Aldrich

4-Chlorophenol

for synthesis

Synonym(s):

4-Chlorophenol, 4-Chloro-1-hydroxybenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(Cl)C6H4OH
CAS Number:
Molecular Weight:
128.56
MDL number:
UNSPSC Code:
12352104
EC Index Number:
203-402-6
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

0.15 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

crystals

bp

220 °C/1013 hPa

transition temp

flash point 121 °C

density

1.26 g/cm3 at 45 °C

storage temp.

2-30°C

SMILES string

Clc1ccc(cc1)O

InChI

1S/C6H5ClO/c7-5-1-3-6(8)4-2-5/h1-4,8H

InChI key

WXNZTHHGJRFXKQ-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

249.8 °F - closed cup

Flash Point(C)

121 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service