Skip to Content
Merck
All Photos(1)

Documents

8.00387

Sigma-Aldrich

Malonic acid

Technipur®, for synthesis

Synonym(s):

1,3-Propanedioic acid, Carboxyacetic acid, Dicarboxymethane, Methanedicarboxylic acid, Propanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2(COOH)2
CAS Number:
Molecular Weight:
104.06
Beilstein:
1751370
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.21

grade

for synthesis

Quality Level

vapor pressure

0.002 hPa ( 25 °C)

mp

132-135 °C (dec.) (lit.)

SMILES string

OC(=O)CC(O)=O

InChI

1S/C3H4O4/c4-2(5)1-3(6)7/h1H2,(H,4,5)(H,6,7)

InChI key

OFOBLEOULBTSOW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Malonic acid is a 1,3-dicarboxylic acid commonly used in the manufacture of dyes, pharmaceuticals, adhesives, agrochemicals, and fragrances, facilitated by the high reactivity of the central methylene group.

Application

Malonic acid can be used as:
  • A complexing agent in the preparation of Cr-P coatings applicable in engineering tools and components.
  • A cross-linker in the synthesis of closed-cell polyurethane foams applicable in the polymer industry.

It can also be introduced into the isotropic conductive adhesive formulation to enhance the electrical properties of electrically conductive adhesives.

Legal Information

Technipur is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

314.6 °F

Flash Point(C)

157 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spectroscopic study of the morphology development of closed-cell polyurethane foam using bio-based malonic acid as chain extender
Zhao W, et al.
Polymer, 193, 122344-122344 (2020)
Electrical property improvement of electrically conductive adhesives through in-situ replacement by short-chain difunctional acids
Li Y, et al.
IEEE Transactions on Components and Packaging Technologies, 29(1), 173-178 (2006)
Preparation and characterization of Cr?P coatings by electrodeposition from trivalent chromium electrolytes using malonic acid as complex
Li B, et al.
Surface and Coatings Technology, 201(6), 2578-2586 (2006)
Malonic Acid and Derivatives
Strittmatter H
Ullmann's Encyclopedia of Industrial Chemistry null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service