Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
26111710
NACRES:
NA.28
Quality Segment
product line
MVP ICON®
manufacturer/tradename
MVP ICON®
application(s)
food and beverages
compatibility
MVP ICON® for 78002 ICON
General description
MVP ICON® Battery Assembly (for 78002 Version Instruments).
SPECIFICATIONS:
Lithium ion cylinder assembly, 3.6V, 2900 mAh, 10.44 Wh
SPECIFICATIONS:
Lithium ion cylinder assembly, 3.6V, 2900 mAh, 10.44 Wh
Application
For use in replacing the existing MVP ICON® Battery used in 78002 Version Instruments.
Packaging
Plast re-sealable bubble wrap bag
Legal Information
BioControl is a registered trademark of BioControl Systems, Inc.
MVP ICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.