Skip to Content
Merck

Skip To

73087BC

Assurance® GDS PickPen® Tips

BioControl, For use with GDS PickPen®, Assurance® GDS PickPen® II Tips, Racked

Synonym(s):

PickPen® Tips

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41122409
NACRES:
NC.81
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

rack of 96 tips

manufacturer/tradename

BioControl Systems 73087

storage condition

room temperature

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

compatibility

for use with PICKPEN®

storage temp.

room temp

General description

GDS PickPen® Tips are the consumable accessories of the GDS PickPen® device. Assurance® GDS is an automated nucleic acid amplification system for the detection of pathogenic organisms in foods, ingredients, and environmental samples. As part of the sample preparation, the proprietary GDS PickPen® is designed to transfer magnetic particles when working with Assurance® GDS pathogen detection kits. The incorporation of the GDS PickPen® in the immunomagnetic separation (IMS) process improves the procedure by concentrating the target pathogen while decreasing the carryover of background microflora from a wide range of food matrices without compromising the sensitivity.

Application

Immunomagnetic Separation (IMS)
The Assurance® GDS PickPen® Tips are used with the PickPen® device to collect and transfer magnetic particles during sample preparation for detection of food pathogens by Assurance® GDS.

Other Notes

One rack of 96 PickPen tips

Legal Information

Assurance is a registered trademark of Merck KGaA, Darmstadt, Germany
PICKPEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
73091BC73085BC79102BC
compatibility

for use with PICKPEN®

compatibility

for use with GDS Rotor-Gene

compatibility

for use with PICKPEN®

compatibility

for use with GDS PICKPEN® PIPETMAX® System

packaging

rack of 96 tips

packaging

-

packaging

bag of 500 tips

packaging

-

storage condition

room temperature

storage condition

-

storage condition

room temperature

storage condition

room temperature

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

application(s)

food and beverages
pathogen testing

application(s)

food and beverages
pathogen testing

application(s)

food and beverages
pathogen testing

application(s)

food and beverages
pathogen testing

technique(s)

PCR: suitable

technique(s)

PCR: suitable

technique(s)

PCR: suitable

technique(s)

PCR: suitable


Still not finding the right product?

Explore all of our products under Assurance® GDS PickPen® Tips




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters