Skip to Content
Merck

71403-M

Rosetta 2(DE3)pLysS Competent Cells - Novagen

Escherichia coli, rod shaped

Synonym(s):

Competent cells, Rosetta DE3 cells

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41106202
NACRES:
NA.81
Biological source:
Escherichia coli
Growth mode:
adherent or suspension
Morphology:
rod shaped
Manufacturer/tradename:
Novagen®
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Rosetta 2(DE3)pLysS Competent Cells - Novagen, Novagen′s Rosetta 2 (pLysS) host strains are BL21 derivatives designed to enhance the expression of eukaryotic proteins that contain codons rarely used in E. coli along with T7 lysozyme activity.

biological source

Escherichia coli

Quality Segment

manufacturer/tradename

Novagen®

storage condition

OK to freeze

growth mode

adherent or suspension

morphology

rod shaped

technique(s)

microbiological culture: suitable

cell transformation

transformation efficiency: >2 × 106 cfu/μg

shipped in

dry ice

storage temp.

−70°C

General description

Genotype: F-ompT hsdSB(rB- mB-) gal dcm (DE3) pLysSRARE2 (CamR)

This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges us to request certain information about you and the establishment where the GMOs are being handled. Click here for Enduser Declaration (EUD) Form.
A common method for transformation of DNA plasmids into E. coli is the use of chemically competent cells. Although competent cells can be prepared in the laboratory, greater efficiency, reproducibility, and convenience are achieved using Novagen prepared competent cells. Novagen competent cells represent the widest selection available for protein expression. Every Novagen competent cell strain is verified for phenotype and purity, and is guaranteed fortransformation efficiency.
T7 expression strains are lysogens of bacteriophageDE3, as indicated by the (DE3). These hosts carry achromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Such strains are suitable for production of protein from target genes cloned in appropriate T7 expression vectors, using IPTG as an inducer.
Rosetta 2 host strains are BL21 derivatives designed to enhance the expression of eukaryotic proteins that contain codons rarely used in E. coli. These strains supply tRNAs for 7 rare codones (AGA, AGG, AUA, CUA, GGA, CCC, and CGG) on a compatible chloramphenicol-resistant plasmid. The tRNA genes are driven by their native promoters.

DE3 indicates that the host is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG.

pLysS strains express T7 lysozyme, which further suppresses basal expression of T7 RNA polymerase prior to induction, thus stabilizing pET recombinants encoding target proteins that affect cell growth and viability.
Novagen′s Rosetta 2 (pLysS) host strains are BL21 derivatives designed to enhance the expression of eukaryotic proteins that contain codons rarely used in E. coli along with T7 lysozyme activity.

Other Notes

•2 × 200 µl or 5 × 200 µl Rosetta 2(DE3)pLysS Competent Cells

•2 × 2 ml or 4 × 2 mlSOC Medium

•10 µlTest Plasmid

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

This product is sold for internal research use only. Any commercial use of this product, its components, and/or any derivatives thereof (including but not limited to proteins produced using the product or its components) (together and hereinafter the “EMD Product”) requires signature of a written commercial use agreement with EMD Millipore Corporation or its successor-in-interest. Commercial use shall include but not be limited to: (1) use of the EMD Product to manufacture products for sale to third parties; (2) use of the EMD Product to provide services, information, or data to third parties in exchange for consideration; (3) use of the EMD Product for therapeutic, diagnostic or prophylactic purposes (including as part of a device, chip, assay or other product); or (4) resale of the EMD Product, whether or not such EMD Product is resold for research use. Nothing contained herein shall be deemed to represent or warrant that additional third party rights are not required for use of the EMD Product. Please direct any questions on these use restrictions to: [email protected].
Toxicity: Multiple Toxicity Values, refer to MSDS (O)


wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library