Skip to Content
Merck

71280

Veggie Peptone - Novagen

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.54
UNSPSC Code:
41106212
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solid

Quality Level

manufacturer/tradename

Novagen®

storage condition

OK to freeze

shipped in

ambient

storage temp.

10-30°C

General description

Veggie Peptone is a certified animal-free media component obtained from papain-digested soymeal that can be used as a direct replacement for tryptone in bacterial growth media. It has been quality tested to ensure proper growth and maintenance of bacterial cells. The Veggie line of products are ideal for applications requiring minimal use of animal-derived materials.

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
71280-307790788055295