Select a Size
About This Item
Quality Level
form
suspension
manufacturer/tradename
Novagen®
storage condition
OK to freeze
technique(s)
transfection: suitable
shipped in
wet ice
storage temp.
2-8°C
General description
- When Insect Genejuice® used in conjunction with Sf9 cell line, raw relative light units (RLUs) from Renilla Luciferase expression with plex is 131,90 Vs. 75,726 when other reagent used with Sf9, thus 1.7 fold difference.
- When Insect Genejuice® used in conjunction with Sf9 cell line, raw relative light units (RLUs) from Renilla Luciferase expression with plex is 131,90 Vs. 11,952 when other reagent used with High Five, thus 11 fold difference.
- When Insect Genejuice® used in conjunction with Sf9 cell line, raw relative light units (RLUs) from Renilla Luciferase expression with plex is 131,90 Vs. 31,897 when Insect Genejuice used with High Five, thus 4.1 fold difference.
Insect GeneJuice Transfection Reagent is provided as a 2 mg/ml suspension in 20 mM MES, 150 mM NaCl, pH 6.2 buffer. One ml is sufficient for 12 or 125 transfections in 10-ml suspension culture flasks or 35-mm plates, respectively.
Legal Information
Disclaimer
Storage Class
12 - Non Combustible Liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Protocols
Related Content
Successful delivery of nucleic acids and protein in eukaryotic cells requires a transfection protocol that maximizes transfection efciency, minimizes cytotoxicity, and results in the desired level of gene expression or silencing. EMD Millipore recognizes that there is not a one-size-ts-all solution to your transfection needs. We provide the breadth of transfection reagents necessary to give you the freedom to design the perfect experiment.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 71259-4 | 04055977270860 |
| 71259-5 | 04055977270877 |
| 71259-3 | 07790788053123 |