Skip to Content
Merck

71077

X-Gal Solution - Novagen

Convenience for blue/white screening

Synonym(s):

X-Gal solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.54
UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

solution

manufacturer/tradename

Novagen®

storage condition

OK to freeze

technique(s)

tissue culture: suitable

shipped in

wet ice

storage temp.

−20°C

General description

X-Gal, or 5-bromo-4-chloro-3-indolyl-β-D-galactopyranoside, is an organic compound commonly used in molecular biology as a chromogenic substrate for the enzyme β-galactosidase, which is encoded by the LacZ gene. When β-galactosidase hydrolyzes X-Gal, it forms galactose and 5,5′-dibromo-4,4′-dichloro-indigo, resulting in the formation of intense blue color colonies. This process is widely used in LacZ activity assays and staining techniques, such as X-gal staining or LacZ staining to distinguish between recombinants (white) and nonrecombinants (blue). X-Gal staining is frequently used in the fields of molecular biology, genetics, and developmental biology to study gene expression and cell line tracing. X-Gal Solution is provided as a convenient 40 mg/ml concentrate in DMSO, ready for dilution into the culture medium or appropriate buffers.

Application

X-Gal Solution - Novagen has been used at 1:40 dilution to detect the expression of β-galactosidase and mutant fusion protein generated by the ApcΔ242 mutation in the tissue sample.

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Irritant (B)


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

188.6 °F

flash_point_c

87 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library