Skip to Content

71013-M

ColiRollers® Plating Beads

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.84
UNSPSC Code:
12352202
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


manufacturer/tradename

Novagen®

Quality Segment

storage condition

OK to freeze

shipped in

ambient

storage temp.

15-25°C

General description

ColiRollers Plating Beads offer a better way to perform a routine procedure-plating bacteria. Typical plating methods use a bent glass rod "hockey stick," which must be immersed in 70% ethanol and flamed between uses, or disposable plastic applicators. Both require vigorous spreading while rotating the plate. These older methods can become extremely laborious and time consuming when processing many samples, and can produce inconsistent results due to differences in pressure applied, length of rotation time, and moisture content of the plates. In addition, an open flame can present a safety hazard. Novagen ColiRollers Plating Beads are specially treated glass beads that do the work for you. Simply pipet the cells on the plate, add 10 to 20 beads, and move the plate back and forth a few times. The rolling action of the beads gently spreads the cells evenly and completely over the agar surface. Entire stacks of plates can be processed at one time. After plating, shake the beads off the plate and the plate is ready for the incubator. Use of ColiRollers beads eliminates the inconsistencies of conventional "hockey stick" methods, and provides even distribution of colonies every time. ColiRollers beads are provided sterile and each package contains enough beads for 60 to 80 plates.

Legal Information

ColiRollers is a trademark of Merck KGaA, Darmstadt, Germany
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity- Standard Handling


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library