Skip to Content

70746

Benzonase® Nuclease

Purity > 90%

Synonym(s):

Endonuclease from Serratia marcescens

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.54
EC Number:
MDL number:
Assay:
>90% (SDS-PAGE)
Biological source:
Serratia marcescens
Recombinant:
expressed in E. coli
Concentration:
25-29 units/μL
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Benzonase® Nuclease, Purity > 90%,

biological source

Serratia marcescens

Quality Segment

recombinant

expressed in E. coli

assay

>90% (SDS-PAGE)

form

buffered aqueous glycerol solution

manufacturer/tradename

Novagen®

storage condition

OK to freeze

concentration

25-29 units/μL

input

general use sample(s)

application(s)

research use

shipped in

wet ice

storage temp.

−20°C

General description

Benzonase® Nuclease is a geneticallyengineered endonuclease from Serratiamarcescens. It degrades all forms of DNA andRNA (single stranded, double stranded, linearand circular) while having no proteolyticactivity. It is effective over a wide range ofconditions and possesses an exceptionallyhigh specific activity. The enzyme completelydigests nucleic acids to 5′-monophosphateterminated oligonucleotides 2 to 5 bases inlength (below the hybridization limit), whichis ideal for removal of nucleic acids fromrecombinant proteins, enabling compliancewith FDA guidelines for nucleic acidcontamination. The ability of Benzonase torapidly hydrolyze nucleic acids makes theenzyme an excellent choice for viscosityreduction to reduce processing time andincrease yields of protein. For example, theenzyme is compatible with BugBuster andPopCulture Protein Extraction Reagentsand can therefore be added along with thesereagents to eliminate viscosity and removenucleic acids from E. coli extracts.

The enzyme consists of two subunits of30 kDa each. It is functional betweenpH 6 and 10 and from 0-42°C and requires1-2 mM Mg2+ for activation. The enzyme is also active in the presence of ionic and non-ionic detergents, reducing agents, PMSF(1 mM), EDTA (1 mM) and urea (relativeactivity depends on specific conditions).Activity is inhibited by >150 mM monovalentcations, >100 mM phosphate, >100 mMammonium sulfate, or >100 mM guanidine HCl.

Benzonase Nuclease is available inultrapure (>99% by SDS-PAGE) and pure(>90%) grades at a standard concentrationof 25-29 U/µl and at a high concentration (HC)of 250 U/µl. Both preparations are free ofdetectable protease and have specific activity> 1 × 106 U/mg protein. The >99% puritygrade is tested for endotoxins and contains<0.25 EU/1000 units. The product is suppliedin 50% glycerol.Store at -20°C.

Total endotoxin:<0.25 EU/1,000 units.Purity: >90% by SDS-PAGE

Application

Used for the removal of nucleic acid from protein samples.

Biochem/physiol Actions

Digests native or heat-denatured DNA and RNA.

Other Notes

One unit is defined as the amount of enzyme that causes a ΔA₂₆₀ of 1.0 in 30 minutes, which corresponds to complete digestion of 37 µg DNA.

Legal Information

Benzonase is a registered trademark of Merck KGaA, Darmstadt, Germany
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Still not finding the right product?

Explore all of our products under Benzonase® Nuclease, Purity > 90%


Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library