Skip to Content
Merck

681685

WP1130

A cell-permeable tyrphostin compound that acts as a partially selective deubiquitinase (DUB) inhibitor and suppresses JAK-Stat signaling pathway.

Synonym(s):

WP1130, Degrasyn

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19H18BrN3O
CAS Number:
Molecular Weight:
384.27
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥97% (HPLC)
Form:
solid
Storage condition:
OK to freeze, protect from light
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

light yellow

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

Brc1nc(ccc1)\C=C(\C(=O)N[C@@H](CCC)c2ccccc2)/C#N

InChI key

LIDOPKHSVQTSJY-VMEIHUARSA-N

General description

A cell-permeable tyrphostin compound that acts as a partially selective deubiquitinase (DUB) inhibitor and suppresses JAK-Stat signaling pathway. Reported to cause accumulation of polyubiquitinated p53, JAK2 and Bcr-Abl, and reduce c-Myc and MCL-1 protein levels in several tumor cells and downregulate Stat3 and NF-κB activities (IC50<1 µM in MCL cells) leading to tumor cell apoptosis (IC50 ~1.2 µM in Z138 cells). Suggested to modify active site cysteine of DUB and inhibit USP5, UCH-L1, USP9x, USP14, and UCH37 activities (≥80% inhibition at 5 µM) without affecting 20S proteasome. Reported to exert stronger antiproliferative and antitumor properties than STAT3 Inhibitor III, WP1066 (Cat. No. 573097) in xenograft mouse models of CML and melanoma.

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Kapuria, V., et al. 2010. Cancer Res.70, 9265.
Pham, L.V., et al. 2010. Mol. Cancer Ther.9, 2026.
Bartholomeusz, G.A, et al. 2007. Cancer Res.67, 3912.

Bartholomeusz, G.A, et al. 2007. Blood109, 3470.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library