Skip to Content
Merck

676484

VEGFR2 Kinase Inhibitor VI, Ki8751

The VEGFR2 Kinase Inhibitor VI, Ki8751, also referenced under CAS 228559-41-9, controls the biological activity of VEGFR2 Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

VEGFR2 Kinase Inhibitor VI, Ki8751, N-(2,4-Difluorophenyl)-Nʹ-(4-(6,7-dimethoxy-4-quinolyloxy)-2-fluorophenyl)urea, Vascular Endothelial Growth Factor Receptor 2 Kinase Inhibitor VI, VEGFR Tyrosine Kinase Inhibitor XXIV

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C24H18F3N3O4
CAS Number:
Molecular Weight:
469.41
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥97% (HPLC)
Form:
solid
Storage condition:
OK to freeze, protect from light
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

off-white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

Fc1c(ccc(c1)F)NC(=O)Nc2c(cc(cc2)Oc3c4c(ncc3)cc(c(c4)OC)OC)F

InChI

1S/C24H18F3N3O4/c1-32-22-11-15-20(12-23(22)33-2)28-8-7-21(15)34-14-4-6-19(17(27)10-14)30-24(31)29-18-5-3-13(25)9-16(18)26/h3-12H,1-2H3,(H2,29,30,31)

InChI key

LFKQSJNCVRGFCC-UHFFFAOYSA-N

General description

A cell-permeable quinolyloxyphenyl-urea compound that acts as a VEGFR-2-selective inhibitior in both cell-based (IC50 = 0.9 nM in VEGF-induced receptor phosphorylation) and cell-free (IC50 = 4.0 nM in kinase reactions) assays. It inhibits c-Kit, PDGFRα, and FGFR-2 only at much higher concentrations (IC50 = 40, 67, and 170 nM, respectively, in cell-based assays), while exhibiting no effect against eight other commonly studied receptor and non-receptor kinases even at concentrations as high as 10 µM. Effectively blocks VEGF-dependent, but not VEGF-independent, cell proliferation in vitro and tumor growth in mice and rats (5 to 20 mg/kg, daily p.o.) in vivo.

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Kubo, K., et al. 2005. J. Med. Chem.48, 1359.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Regulatory Review (Z)


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library