Skip to Content
Merck
All Photos(1)

Key Documents

648464

Millipore

Triton X-100, Hydrogenated, PROTEIN GRADE® Detergent, 10% Solution, Sterile-Filtered

Non-ionic detergent used for the solubilization of membrane-bound proteins.

Synonym(s):

Octylphenoxypolyethoxyethanol, Hydrogenated

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161902
NACRES:
NA.25

description

Merck USA index - 14, 6761

Quality Level

form

solution

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤200 μM Aldehyde
≤200 μM peroxide

color

clear colorless

conductivity

≤50 μmho

CMC

250 μM

shipped in

ambient

storage temp.

2-8°C

General description

Non–ionic detergent used for the solubilization of membrane-bound proteins. This purified detergent is advantageous for use with sensitive, membrane-bound proteins, overcoming the problems associated with aldehyde and peroxide impurities in polyoxyethylene detergents. Unlike its parent compound, Hydrogenated TRITON X-100 Detergent lacks UV absorbance and fluorescence, which cause interference in spectophotometric monitoring and assays.
Nonionic detergent used for the solubilization of membrane-bound proteins. This purified detergent is advantageous for use with sensitive, membrane-bound proteins, overcoming the problems associated with aldehyde and peroxide impurities in polyoxyethylene detergents. Unlike its parent compound, Hydrogenated TRITON X-100 lacks UV absorbance and fluorescence, which cause interference in spectophotometric monitoring and assays. Has a critical micelle concentration (CMC) of 250 µM.

Application


  • Intrinsic Lipid Curvature and Bilayer Elasticity as Regulators of Channel Function: A Comparative Single-Molecule Study.: This study investigates how intrinsic lipid curvature and bilayer elasticity affect channel function using single-molecule techniques. Triton X-100, Hydrogenated, PROTEIN Detergent, 10% Solution, Sterile-Filtered is used to solubilize membrane proteins, enabling detailed analysis of lipid-protein interactions and their impact on membrane dynamics (Ashrafuzzaman et al., 2024).

  • Preparing for another Ebola Outbreak: The impact of viral inactivation methods on commonly measured biochemistry analytes in plasma and urine.: This study examines the effect of viral inactivation methods on biochemistry analytes in plasma and urine, using Triton X-100 for viral inactivation. The findings are crucial for ensuring accurate biochemical measurements during viral outbreaks (Zhou et al., 2024).


Packaging

Packaged under inert gas

Warning

Toxicity: Harmful (C)

Physical form

10% aqueous solution.

Other Notes

Fernandez, J., et al. 1992. Anal. Biochem.201, 255.
Tiller, G.E., et al. 1984. Anal. Biochem.141, 262.
Ashani, Y. and Cantravas, G.N. 1980. Anal. Biochem.109, 55.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
PROTEIN GRADE is a registered trademark of Merck KGaA, Darmstadt, Germany
Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ping Wang et al.
Current protocols, 1(8), e174-e174 (2021-08-06)
Chromatin Interaction Analysis Using Paired-End Tag Sequencing (ChIA-PET) is an established method to map protein-mediated chromatin interactions. A limitation, however, is that it requires a hundred million cells per experiment, which hampers its broad application in biomedical research, particularly in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service