Skip to Content
Merck

596200

Thioflavin T

A cell-permeable benzothiazole dye that exhibits enhanced fluorescence upon binding to amyloid fibrils.

Synonym(s):

Thioflavin T, ThT, Thioflavine T, Basic Yellow 1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C17H19ClN2S
CAS Number:
Molecular Weight:
318.86
UNSPSC Code:
12171500
NACRES:
NA.25
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

yellow

solubility

water: 5 mg/mL

fluorescence

λex ~385 nm (free), λex ~437 nm, λem ~445 nm (free), λem ~485 nm

shipped in

ambient

storage temp.

15-25°C

SMILES string

S1c2c(ccc(c2)C)N(C1=C3C=CC(=[N+](C)C)C=C3)C.[Cl-]

InChI

1S/C17H19N2S.ClH/c1-12-5-10-15-16(11-12)20-17(19(15)4)13-6-8-14(9-7-13)18(2)3;/h5-11H,1-4H3;1H/q+1;/p-1

InChI key

JADVWWSKYZXRGX-UHFFFAOYSA-M

General description

A cell-permeable benzothiazole dye that exhibits enhanced fluorescence upon binding to amyloid fibrils. Useful in monitoring stacked β sheet aggregates. Associates with aggregated fibrils of synthetic β/A4-derived peptides β(1-28) and β(1-40), resulting in a shift in the excitation maximum and an enhanced emission. Excitation max.: ~385 nm (free); ~437 nm (bound); emission max.: ~445 nm (free); ~485 nm (bound).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Exhibits enhanced fluorescence upon binding to amyloid fibrils
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Tjernberg, L.O., et al. 1999. J. Biol. Chem.274, 12619.
Naiki, H. and Nakakuki, K. 1996. Lab. Invest.74, 374.
LeVine, H. 1993. Protein Sci.2, 404.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Still not finding the right product?

Explore all of our products under Thioflavin T


signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Sens. 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
596200-500MG04055977189131