Skip to Content
Merck

5650

L-Serine

Synonym(s):

L-Serine, Ser

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C3H7NO3
CAS Number:
Molecular Weight:
105.09
UNSPSC Code:
12352209
NACRES:
NA.25
MDL number:
eCl@ss:
32160406
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

RTECS - VT8100000
Merck USA index - 14, 8460

Quality Level

assay

≥99% (titration)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: 5 mg/mL

cation traces

heavy metals: ≤15 ppm

shipped in

ambient

storage temp.

15-25°C

SMILES string

[N+H3]C(CO)C(=O)[O-]

InChI

1S/C3H7NO3/c4-2(1-5)3(6)7/h2,5H,1,4H2,(H,6,7)

InChI key

MTCFGRXMJLQNBG-UHFFFAOYSA-N

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Still not finding the right product?

Explore all of our products under L-Serine


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
M4001-1G04061826060490
M4001-100MG04061832417356
5650-100GM04055977267563