Skip to Content

539149

Proteasome Substrate VI, Fluorogenic

A fluorogenic substrate probe useful for assaying trypsin-like activity of proteasome.

Synonym(s):

Proteasome Substrate VI, Fluorogenic, Z-ARR-AMC, 2HCl

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C33H44N10O7 · 2HCl
CAS Number:
Molecular Weight:
765.69
MDL number:
NACRES:
NA.77
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
solid
Storage condition:
OK to freeze, protect from light
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

white to off-white

solubility

DMSO: 200 mg/mL, 80% acetic acid: 50 mg/mL, methanol: soluble

fluorescence

λex ~380 nm, λem ~460 nm

shipped in

ambient

storage temp.

−20°C

General description

A fluorogenic substrate for assaying trypsin-like activity of proteasome. Also serves as a substrate for cathepsin B. Excitation max: 360-380 nm; emission max.: 430-460 nm.
A fluorogenic substrate probe useful for assaying trypsin-like activity of proteasome. Also reported to be a specific substrate for cathepsin B.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Proteasome
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Physical form

Supplied as hydrochloride salt.

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for 3 months at -20°C

Other Notes

Geier, E., et al. 1999. Science283, 978.
Dick, T.P., et al. 1998. J. Biol. Chem. 273, 25637.
Z-Ala-Arg-Arg-AMC

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library