Skip to Content
Merck
All Photos(1)

Key Documents

539132

Millipore

Protease Inhibitor Cocktail II

lyophilized, for the inhibition of aspartic, cysteine, serine, metalloproteases and aminopeptidases. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

Protease inhibitor cocktail

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.54
form:
lyophilized
solubility:
water: soluble
storage temp.:
−20°C

Product Name

Protease Inhibitor Cocktail Set II, The Protease Inhibitor Cocktail Set II controls the activity of Protease. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Quality Level

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

solubility

water: soluble

shipped in

wet ice

storage temp.

−20°C

General description

A cocktail of five protease inhibitors with broad specificity for the inhibition of aspartic, cysteine, serine, and metalloproteases as well as aminopeptidases. This cocktail is recommended for use with bacterial cell extracts.
Note: 1 set = 1 vial of lyophilized protease inhibitor cocktail and 1 vial DMSO, 1 ml; 5 sets = 5 vials of lyophilized protease inhibitor cocktail and 5 vials of DMSO, 1 ml.
Protease Inhibitor Cocktail Set II is a specially formulated cocktail of five protease inhibitors with broad specificity for the inhibition of aspartic, cysteine, and serine and metalloproteases as well as aminopeptidases. This cocktail is recommended for use with bacterial cell extracts.

1 set (1 vial of protease inhibitor cocktail plus 1 vial of 1.0 ml DMSO)

5 sets (5 vials of protease inhibitor cocktail plus 5 vials of 1.0 ml DMSO)

Specificity

Inhibits a broad spectrum of serine proteases, cysteine proteases, aspartic proteases, and metalloproteases, as well as aminopeptidases.

Application

Protease Inhibitor Cocktail Set II, contains five protease inhibitors for aspartic, cysteine, serine, metalloproteases, and aminopeptidases. Recommended bacterial cell extracts.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Aspartic, cysteine, serine, and metalloproteases as well as aminopeptidases

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze at -20°C. The reconstituted material is stable for up to 6 months at -20°C.
This product is provided as a lyophilized solid, ready for reconstitution. For the researcher′s convenience, this protease inhibitor cocktail is provided as either a single vial plus one vial of 1.0 ml DMSO or as a set of 5 vials plus 5 vials of 1.0 ml DMSO. Reconstitute each vial of lyophilized protease inhibitor cocktail with 1 ml of DMSO first. Mix gently. Then add 4 ml of water. The solution will appear turbid.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Serena Vella et al.
The international journal of biochemistry & cell biology, 76, 1-11 (2016-05-02)
Cardiac progenitors, such as cardiospheres and cardiosphere-derived cells, represent an attractive cell source for cardiac regeneration. The PIWI-interacting RNAs, piRNAs, are an intriguing class of small non-coding RNAs, implicated in the regulation of epigenetic state, maintenance of genomic integrity and
Tom40 in cholesterol transport.
Bose, et al.
iScience, 26, 106386-106386 (2023)
Hong Lv et al.
iScience, 27(6), 109667-109667 (2024-07-05)
Our work aimed to investigate the interactive roles of transforming growth factor β1 (TGF-β1), ubiquitin-specific-processing protease 7 (USP7), and Yes-associated protein (YAP) in ferroptosis during sepsis-secondary acute lung injury (ALI). Our study demonstrated that ferroptosis was aggravated by TGF-β1 in
Andrew T Crombie
Environmental microbiology (2021-07-29)
The biological importance of lanthanides has only recently been identified, initially as the active site metal of the alternative methanol dehydrogenase (MDH) Xox-MDH. So far, the effect of lanthanide (Ln) has only been studied in relatively few organisms. This work
Evan C Ray et al.
American journal of physiology. Renal physiology, 321(6), F705-F714 (2021-10-12)
The epithelial Na+ channel (ENaC) promotes the absorption of Na+ in the aldosterone-sensitive distal nephron, colon, and respiratory epithelia. Deletion of genes encoding subunits of ENaC results in early postnatal mortality. Here, we present the initial characterization of a mouse

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service