Skip to Content
Merck
All Photos(1)

Key Documents

5.08839

Sigma-Aldrich

BIBR1532

≥98% (HPLC), solid, telomerase inhibitor, Calbiochem®

Synonym(s):

Telomerase Inhibitor X, BIBR1532, BIBR 1532, BIBR-1532

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H17NO3
CAS Number:
Molecular Weight:
331.36
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Telomerase Inhibitor X, BIBR1532,

Assay

≥98% (HPLC)

Quality Level

form

solid

potency

93 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

SMILES string

N(c3c(cccc3)C(=O)O)C(=O)\C=C(/C)\c1cc2c(cc1)cccc2

InChI

1S/C21H17NO3/c1-14(16-11-10-15-6-2-3-7-17(15)13-16)12-20(23)22-19-9-5-4-8-18(19)21(24)25/h2-13H,1H3,(H,22,23)(H,24,25)/b14-12+

InChI key

PGFQXGLPJUCTOI-WYMLVPIESA-N

General description

A cell-permeable benzoic acid based compound that acts as a selective, highly potent, non-nucleotide competitive inhibitor of the catalytic activity of telomerase (IC50 = 93 nM in HeLa cells nuclear extracts). Also shown to be effective against recombinant, affinity purified telomerase. Blocks the growth and proliferation of HeLa cells (~ 20 µM) by inducing apoptosis. The inhibition appears to be more prominent in TEL patch mutant cell lines. Causes a continuous erosion of telomeres in multiple human cancer cell lines. Does not affect the activity of DNA and RNA polymerases in any significant manner.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Teomerase
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Damm, K., et al. 2001. The. EMBO J.20, 6958.
Pascolo, E., et al. 2002. J. Biol. Chem.277,15566.

Nakashima, M., et al. 2013. J. Biol. Chem.288, 33171.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service