Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C29H33ClN2O2 · xHCl
CAS Number:
Molecular Weight:
477.04 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥99% (HPLC)
Form:
solid
Storage condition:
OK to freeze, protect from light
assay
≥99% (HPLC)
Quality Segment
form
solid
potency
2 nM Ki
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze, protect from light
color
white
solubility
DMSO: 50 mM, ethanol: 50 mM
storage temp.
2-8°C
SMILES string
[Cl-].Clc1ccc(cc1)C2(CC[N+H](CC2)CCC(c4ccccc4)(c3ccccc3)C(=O)N(C)C)O
InChI
1S/C29H33ClN2O2.ClH/c1-31(2)27(33)29(24-9-5-3-6-10-24,25-11-7-4-8-12-25)19-22-32-20-17-28(34,18-21-32)23-13-15-26(30)16-14-23;/h3-16,34H,17-22H2,1-2H3;1H
InChI key
PGYPOBZJRVSMDS-UHFFFAOYSA-N
General description
A very potent and highly selective peripheral resctricted µ-opioid receptor agonist (Ki = 2 nM). Often used as an antidiarrheal, antipruritic, and antihyperalgesic agent. In addition, shown to improve defective post-receptor insulin signaling in skeletal muscle cells and enhance insulin sensitivity in rats.
Biochem/physiol Actions
Primary Target
u opioid receptor
u opioid receptor
Preparation Note
Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.
Warming is required for complete solubilization in ethanol.
Other Notes
Cheng, K., et al. 2013. Pharmaco. Ther.139, 334.
Di Bosco, M., et al. 2008. Chem. Biol. Drug Des.71, 328.
DeHaven-Hudkins, D., et al. 1999. J. Pharmacol. Exp. Ther.289, 494.
Mackerer, C., et al. 1976. J. Pharmacol. Exp. Ther.199, 131.
Di Bosco, M., et al. 2008. Chem. Biol. Drug Des.71, 328.
DeHaven-Hudkins, D., et al. 1999. J. Pharmacol. Exp. Ther.289, 494.
Mackerer, C., et al. 1976. J. Pharmacol. Exp. Ther.199, 131.
Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Toxicity: Toxic (F)
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 3 Oral
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
