Skip to Content
Merck
All Photos(1)

Key Documents

481407

Sigma-Aldrich

NF-κB Activation Inhibitor

InSolution, ≥98%

Synonym(s):

InSolution NF-κB Activation Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H20N4O
Molecular Weight:
356.42
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

shipped in

wet ice

storage temp.

2-8°C

General description

A cell-permeable quinazoline compound that acts as a potent inhibitor of NF-κB transcriptional activation (IC50 = 11 nM in Jurkat cells) and LPS-induced TNF-α production (IC50 = 7 nM in murine splenocytes). Does not exhibit cellular cytotoxicity even at concentrations as high as 10 µM. Reported to exhibit anti-inflammatory effects on carregeenin-induced paw edema in rats.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
NF-κB transcriptional activation
Product does not compete with ATP.
Reversible: no
Target IC50: 11 nM against NF-κB transcriptional activation in Jurkat cells; 7 nM against LPS-induced TNF-α production in murine splenocytes

Packaging

Packaged under inert gas

Warning

Toxicity: Toxic (F)

Physical form

A 10 mM (1 mg/281 µl) solution of NF-κB Activation Inhibitor (Cat. No. 481406) in DMSO.

Reconstitution

Following initial use, aliquot and refrigerate (4°C).

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Tobe, M., et al. 2003. Bioorg. Med. Chem.11, 383.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - closed cup - (Dimethylsulfoxide)

Flash Point(C)

87 °C - closed cup - (Dimethylsulfoxide)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masanori Tobe et al.
Bioorganic & medicinal chemistry, 11(3), 383-391 (2003-01-09)
We disclose here a new structural class of low-molecular-weight inhibitors of NF-kappa B activation that were designed and synthesized by starting from quinazoline derivative 6a. Structure-activity relationship (SAR) studies based on 6a elucidated the structural requirements essential for the inhibitory
Differential regulation of transcription factor T-bet induction during NK cell development and T helper-1 cell differentiation.
Fang, et al.
Immunity, 55, 639-655 (2023)
Fatmah A Mansour et al.
Oncoimmunology, 9(1), 1729299-1729299 (2020-04-22)
The T-cell inhibitory molecule PD-L1 is expressed on a fraction of breast cancer cells. The distribution of PD-L1 on the different subpopulations of breast cancer cells is not well-defined. Our aim was to study the expression level of PD-L1 on
Eunbi Ko et al.
Experimental dermatology, 24(12), 936-941 (2015-07-16)
House dust mites (HDMs) are known to trigger chronic inflammation through Toll-like receptors (TLRs) and their signalling cascades. In this study, we found that TLR2 ligation by HDMs induced the activation of dual oxidase 2 (Duox2) and nuclear factor-κB (NF-κB)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service