Select a Size
About This Item
Product Name
Mitochondrial Division Inhibitor, mdivi-1, The Mitochondrial Division Inhibitor, mdivi-1, also referenced under CAS 338967-87-6, controls the biological activity of yeast Dnm1 and mammalian Drp1.
Quality Segment
assay
≥95% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze, protect from light
color
white
solubility
DMSO: 10 mg/mL
shipped in
wet ice
storage temp.
−20°C
SMILES string
Sc1[n]([c](c3c(n1)cccc3)=O)c2c(cc(c(c2)OC)Cl)Cl
InChI
1S/C15H10Cl2N2O2S/c1-21-13-7-12(9(16)6-10(13)17)19-14(20)8-4-2-3-5-11(8)18-15(19)22/h2-7H,1H3,(H,18,22)
InChI key
NZJKEVWTYMOYOR-UHFFFAOYSA-N
General description
Packaging
Other Notes
Legal Information
Disclaimer
Still not finding the right product?
Explore all of our products under Mitochondrial Division Inhibitor, mdivi-1
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.