Skip to Content

457250

Methylene Blue

guanylate cyclase inhibitor, solid

Synonym(s):

Methylthioninium chloride, 3,7- bis(Dimethylamino)phenothiazin-5-ium Chloride, 3,7-bis(Dimethylamino)phenothiazin-5-ium Chloride, Methylene blue

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C16H18ClN3S
CAS Number:
Molecular Weight:
319.85
UNSPSC Code:
12171500
NACRES:
NA.77
MDL number:
Form:
solid
Storage condition:
OK to freeze
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Methylene Blue, Inhibitor of soluble guanylate cyclase.

Quality Segment

description

Merck USA index - 14, 6060

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

dark green

solubility

water: 4 mg/mL

shipped in

ambient

storage temp.

10-30°C

SMILES string

[s]1c2c(nc3c1cc(cc3)N(C)C)cc[c](c2)=[N+](C)C.[Cl-]

InChI

1S/C16H18N3S.ClH/c1-18(2)11-5-7-13-15(9-11)20-16-10-12(19(3)4)6-8-14(16)17-13;/h5-10H,1-4H3;1H/q+1;/p-1

InChI key

CXKWCBBOMKCUKX-UHFFFAOYSA-M

General description

Inhibitor of soluble guanylate cyclase. Organic spin-trap agent that forms stable adducts with oxygen free radicals in solution; biological staining agent. Dye content: ≥85%.

Other Notes

Kannan, M.S., and Johnson, D.E. 1995. Life Sci.56, 2229.
Hedrum, A., et al. 1994. Biotechniques17, 118.
Lim, C.S., and Hunt, C.A. 1994. Biotechniques17, 626.
Yezzi, M.J., et al. 1994. J. Immunol.152, 3064.
Mayer, B., et al. 1993. Biochem. Pharmacol.45, 367.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Harmful (C)


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library