Skip to Content
Merck

4403-M

Lysozyme, Chicken Egg White

Native, chicken egg white lysozyme. Inhibits the biological activity of lipopolysaccharides from periodontopathic bacteria.

Synonym(s):

Lysozyme, Chicken Egg White, Muramidase, N-Acetylmuramyl Hydrolase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
EC Number:
UNSPSC Code:
12352202
NACRES:
NA.77
MDL number:
Specific activity:
≥20,000 Shugar units/mg dry wt
Biological source:
chicken egg white
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

chicken egg white

description

RTECS - OL5989000
Merck USA index - 14, 5640

form

lyophilized solid

specific activity

≥20,000 Shugar units/mg dry wt

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

technique(s)

cell based assay: suitable

color

off-white

solubility

cold water: 5 mg/mL, aqueous buffer: soluble

suitability

suitable for cell lysis

application(s)

cell analysis

storage temp.

−20°C

General description

Native, chicken egg white lysozyme. Hydrolyzes β1,4 linkages between N-acetylmuramic acid (2-acetamido-2-deoxy-3-O-[1carboxymethyl]-glucopyranose) and N-acetyl-glucosamine residues in chitin. Also hydrolyzes β1,4 linkages between N-acetylmuramic acid and N-acetyl-glucosamine residues in peptidoglycans. Inhibits the biological activity of lipopolysaccharides from periodontopathic bacteria.

Preparation Note

Following reconstitution, aliquot and freeze (-20°C) for long term storage or refrigerate (4°C) for short term storage. Stock solutions are stable for up to 3 days at 4°C or for up to 1 year at -20°C.

Other Notes

Ito, H.O., et al. 1997. J. Periodontal Res. 32, 295.
One Shugar unit is defined as the amount of enzyme that will cause a decrease in absorbance of 0.001 at 450 nm per min due to lysis of Micrococcus luteus at 25°C, pH 6.2.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library