Skip to Content

435800

Licochalcone-A, Synthetic

≥96% (HPLC), estrogenic flavonoid, solid

Synonym(s):

Licochalcone-A, Synthetic, (E)-3-[5-(1,1-Dimethyl-2-propenyl)-4-hydroxy-2-methoxyphenyl]-1-(4-hydroxyphenyl)-2-propen-1-one

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C21H22O4
CAS Number:
Molecular Weight:
338.40
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥96% (HPLC)
Form:
solid
Storage condition:
OK to freeze, protect from light
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Licochalcone-A, Synthetic, An estrogenic flavonoid with anti-tumor and antiparasitic properties.

Quality Segment

assay

≥96% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

yellow

solubility

DMSO: 10 mg/mL, ethanol: soluble

shipped in

ambient

storage temp.

−20°C

SMILES string

O(C)c1c(cc(c(c1)O)C(C)(C)C=C)\C=C\C(=O)c2ccc(cc2)O

InChI

1S/C21H22O4/c1-5-21(2,3)17-12-15(20(25-4)13-19(17)24)8-11-18(23)14-6-9-16(22)10-7-14/h5-13,22,24H,1H2,2-4H3/b11-8+

InChI key

KAZSKMJFUPEHHW-DHZHZOJOSA-N

General description

An estrogenic flavanoid with anti-tumor and anti-parasitic properties. Shown to reduce Bcl-2 protein expression and induce apoptosis in several human cancer cell lines. Also induces cell differentiation and enhances the effect of paclitaxel and vinblastine chemotherapy. Reported to interfere with parasite mitochondrial electron transport chain and energy metabolism.
An estrogenic flavonoid with anti-tumor and antiparasitic properties. Shown to reduce Bcl-2 protein expression and induce apoptosis in several human cancer cell lines. Also induces cell differentiation, exhibits anti-tumor activity, and enhances the effect of Paclitaxel (Cat. No. 580555) and Vinblastine (Cat. No. 677175) chemotherapy. Reported to interfere with parasite mitochondrial electron transport chain and energy metabolism.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Anti-tumor and antiparasitic properties
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution aliquot and freeze at -20°C. Stock solutions are stable for up to 6 months at -20°C.
Further dilute 1:10 with aqueous buffer or medium just prior to use.

Other Notes

Rafi, M.M., et al. 2000. Anticancer Res. 20, 2653.
Zhai, L., et al. 1999. J. Antimicrob. Chemother. 43, 793.
Haraguchi, H., et al. 1998. Phytochemistry 48, 125.
Park, E.J., et al. 1998. Planta Med. 64, 464.
Chen, M., et al. 1994. Antimicrob. Agents Chemother. 38, 1470.
Chen, M., et al. 1993. Antimicrob. Agents. Chemother. 37, 2550.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Harmful (C)


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library