401353
Goat Anti-Rabbit IgG, H & L Chain Specific Peroxidase Conjugate
liquid, Calbiochem®
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
goat
Quality Level
antibody form
affinity isolated antibody
antibody product type
secondary antibodies
clone
polyclonal
form
liquid
contains
0.02% Thimerosal as preservative
manufacturer/tradename
Calbiochem®
storage condition
do not freeze
isotype
IgG
shipped in
wet ice
storage temp.
2-8°C
target post-translational modification
unmodified
General description
Adsorbed against bovine, horse, and human serum proteins.
Immunoaffinity purified goat polyclonal antibody adsorbed against normal human, bovine, and horse serum proteins and conjugated to horseradish peroxidase (HRP). Recognizes rabbit IgG, heavy and light chains.
This Goat Anti-Rabbit IgG, H & L Chain Specific Peroxidase Conjugate is validated for use in Enzyme Immunoassay, Immunoelectrophoresis for the detection of Rabbit IgG, H & L Chain Specific.
Application
Enzyme Immunoassay (1:10,000)
Immunoelectrophoresis (see comments)
Immunoelectrophoresis (see comments)
Warning
Toxicity: Standard Handling (A)
Physical form
In 10 mM PBS, 0.5 mM EDTA, 10% mannose, 2% sucrose, 1% BSA, pH 7.6.
Other Notes
Specificity for rabbit IgG, heavy and light chains, determined by ELISA against normal rabbit serum. Cross-reactivity with normal bovine, horse, and human sera is <2% by direct solid phase immunoassay. Variables associated with assay conditions will dictate the proper working dilution.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
To comply with ban of sale of mercury-added products required by The Interstate Mercury Education and Reduction Clearinghouse (IMERC), this product is prohibited to be sold in the following US states: Rhode Island and Connecticut.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Anticancer research, 39(12), 6711-6722 (2019-12-08)
Autophagy is a cellular mechanism that recycles cellular components to maintain homeostasis. To investigate the clinical implication of autophagy in gastric cancer, the autophagy markers with autophagosome formation, LC3B and selective autophagy substrate p62/SQSTM1 (P62) were validated. LC3B and p62
Translational oncology, 11(1), 109-115 (2017-12-09)
Partitioning defective (Par) proteins regulate cell polarity and differentiation. Par3, Par6β, and protein kinase Cζ (PKCζ), which are PAR complex members, have been shown to be associated with oncogenesis and progression. Herein, we report the expression pattern and clinical relevance
Infection and immunity, 77(11), 4953-4965 (2009-08-26)
The intracellular bacterium Francisella tularensis survives and replicates within macrophages, ultimately killing the host cell. Resolution of infection requires the development of adaptive immunity through presentation of F. tularensis antigens to CD4+ and CD8+ T cells. We have previously established
Nutrients, 12(2) (2020-02-13)
Ginsenoside Rg3, one of the major components in Panax ginseng, has been reported to possess several therapeutic effects including anti-obesity properties. However, its effect on the browning of mature white adipocytes as well as the underlying mechanism remains poorly understood.
Clinical epigenetics, 11(1), 102-102 (2019-07-14)
Apabetalone (RVX-208) is a bromodomain and extraterminal protein inhibitor (BETi) that in phase II trials reduced the relative risk (RR) of major adverse cardiac events (MACE) in patients with cardiovascular disease (CVD) by 44% and in diabetic CVD patients by
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service