Skip to Content
Merck
All Photos(2)

Key Documents

375368

Millipore

HEPES

≥99% dry basis (titration), 1M solution, amniosulfonic acid buffer

Synonym(s):

HEPES, 4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H18N2O4S
CAS Number:
Molecular Weight:
238.30
Beilstein:
883043
MDL number:
UNSPSC Code:
12161700
NACRES:
NA.25

Product Name

HEPES, ULTROL® Grade, ≥99% dry basis (titration), 1M zwitterionic N-substituted aminosulfonic acid buffer

Quality Level

description

Merck USA index - 144654

Assay

≥99% dry basis (titration)

form

solution

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

clear colorless

useful pH range

6.8-8.2

pKa (25 °C)

7.5

shipped in

ambient

storage temp.

15-25°C

SMILES string

OCCN1CCN(CC1)CCS(O)(=O)=O

InChI

1S/C8H18N2O4S/c11-7-5-9-1-3-10(4-2-9)6-8-15(12,13)14/h11H,1-8H2,(H,12,13,14)

InChI key

JKMHFZQWWAIEOD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A zwitterionic N-substituted aminosulfonic acid buffer absorbance: <0.05 at 260 nm, 1 M, H2O; pKa: 7.55 at 20°C). Active in the pH range of 6.0 to 8.5.

Warning

Toxicity: Standard Handling (A)

Physical form

1.0 M solution of Cat. No. 391338. Sterile-filtered.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTROL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service