Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Assay:
≥99% (TLC)
Form:
solid
Quality level:
Storage condition:
OK to freeze, desiccated (hygroscopic)
Product Name
2-Guanidinoethylmercaptosuccinic Acid, Potent inhibitor of a carboxypeptidase B-like processing enzyme referred to as enkephalin convertase (Ki = 8.8 nM).
Quality Level
assay
≥99% (TLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze, desiccated (hygroscopic)
color
white
solubility
water: soluble
shipped in
ambient
storage temp.
2-8°C
SMILES string
S(C(CC(=O)O)C(=O)O)CCN=C(N)N
InChI
1S/C7H13N3O4S/c8-7(9)10-1-2-15-4(6(13)14)3-5(11)12/h4H,1-3H2,(H,11,12)(H,13,14)(H4,8,9,10)
InChI key
VKVCLXDFOQQABP-UHFFFAOYSA-N
General description
Potent inhibitor of a carboxypeptidase B-like processing enzyme referred to as enkephalin convertase (Ki = 8.8 nM). Ideal for use in affinity chromatography of the enzyme.
Potent inhibitor of carboxypeptidase B-like processing enzyme referred to as enkephalin convertase (Ki = 8.8 nM). Ideal for use in affinity chromatography of the enzyme.
Biochem/physiol Actions
Cell permeable: no
Primary Target
Enkephalin convertase
Enkephalin convertase
Product does not compete with ATP.
Reversible: no
Target Ki: 8.8 nM for enkephalin convertase
Other Notes
Bommer, M., et al. 1989. Brain Res. 492, 305.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Toxicity: Standard Handling (A)
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 369334-5MG | 04055977191615 |