Skip to Content
Merck
All Photos(1)

Key Documents

345862

Sigma-Aldrich

Golgicide A

A cell-permeable quinoline compound that selectively targets ArfGEF GBF1, but not BIG1/2, and attenuates GBF1-mediated cellular vesicle traffickings in a reversible manner.

Synonym(s):

Golgicide A, 6,8-Difluoro-4-pyridin-3-yl-3a,4,5,9b-tetrahydro-3H-cyclopenta[c]quinoline, GCA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H14F2N2
CAS Number:
Molecular Weight:
284.30
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

Fc1c2c(cc(c1)F)[C@@H]3[C@H](C(N2)c4cnccc4)CC=C3

InChI

1S/C17H14F2N2/c18-11-7-14-12-4-1-5-13(12)16(10-3-2-6-20-9-10)21-17(14)15(19)8-11/h1-4,6-9,12-13,16,21H,5H2/t12-,13+,16?/m0/s1

InChI key

NJZHEQOUHLZCOX-FTLRAWMYSA-N

General description

A cell-permeable quinoline compound that selectively targets ArfGEF GBF1, but not BIG1/2, and attenuates GBF1-mediated cellular vesicle traffickings in a reversible manner. Phenotypic comparisons in Vero (African Green Monkey Kidney epithelial) cultures reveal that selective blockage of GBF1 function by GCA (10 µM) or by FGB1-E794K dominant-inactive expression affects medial- and cis-Golgi similarly to that seen with BFA (Cat. No. 203729), however, the non-GBF1-specific BFA (10 µg/ml or 35.6 µM) causes additional GBF1-independent morphologic effects on TGN, notably the rapid dispersal of Golgi-associated coat proteins AP-1 and GGA3, which are not seen with GCA treatment. Endocytic/retrograde transport studies using GCA establishes that GBF1 function is not required for the transport of bacterial toxins to the endosomes, however the retrograde transport of StxB (shiga toxin B subunit) from endosomes to TGN and Golgi is GBF1-dependent (100% inhibition of StxB sulfation at 10 µM), accounting for GCA′s (10 µM) ability to completely prevent Vera cells from Stx-induced (up to 100 ng/ml) inhibition of cellular protein synthesis.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Saenz, J.B., et al. 2009. Nat. Chem. Biol.5, 157.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service