Skip to Content
Merck

324759

Elastase Inhibitor IV

The Elastase Inhibitor IV, also referenced under CAS 127373-66-4, controls the biological activity of Elastase. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

Elastase Inhibitor IV, N-( o-( p-Pivaloyloxybenzene)sulfonylaminobenzoyl)glycine, N-(2-(4-(2,2-Dimethylpropionyloxy)phenylsulfonylamino)benzoyl)aminoacetic acid, N-(o-(p-Pivaloyloxybenzene)sulfonylaminobenzoyl)glycine, N-(2-(4-(2,2-Dimethylpropionyloxy)phenylsulfonylamino)benzoyl)aminoacetic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H22N2O7S
CAS Number:
Molecular Weight:
434.46
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥95% (HPLC)
Form:
solid
Quality level:
Storage condition:
OK to freeze, protect from light
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

white to off-white

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(Nc2c(cccc2)C(=O)NCC(=O)O)c1ccc(cc1)OC(=O)C(C)(C)C

InChI

1S/C20H22N2O7S/c1-20(2,3)19(26)29-13-8-10-14(11-9-13)30(27,28)22-16-7-5-4-6-15(16)18(25)21-12-17(23)24/h4-11,22H,12H2,1-3H3,(H,21,25)(H,23,24)

InChI key

BTGNGJJLZOIYID-UHFFFAOYSA-N

General description

A cell-permeable sulfanilide compound that acts as a potent, substrate-competitive, and highly specific inhibitor of neutrophil elastase (IC50 = 19-49 nM in rat, rabbit, hamster, human, and mouse leukocyte elastase). Displays >100-fold greater selectivity over pancreas elastase (IC50 = 5.6 µM). Does not inhibit trypsin, thrombin, plasmin, kallikrein, chymotrypsin, and cathepsin G even at concentrations as high as 100 µM. Effectively suppresses human neutrophil elastase-induced lung hemorrhage and skin capillary permeability in vivo. Also reported to inhibit mast cell Stat6-protease.
A cell-permeable, potent, substrate-competitive, and highly specific inhibitor of neutrophil elastase (IC50 = 19-49 nM using rat, rabbit, hamster, human, and mouse leukocyte elastase). Displays about 100-fold greater selectivity over pancreatic elastase (IC50 = 5.6 µM) and is inactive against trypsin, thrombin, plasmin, kallikrein, chymotrypsin, and cathepsin G even at concentrations as high as 100 µM. Effectively suppresses human neutrophil elastase-induced lung hemorrhage and skin capillary permeability in vivo. Also reported to inhibit mast cell Stat6-protease.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Neutrophil elastase
Product does not compete with ATP.
Reversible: no
Target IC50: 19-49 nM in rat, rabbit, hamster, human, and mouse leukocyte elastase

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Suzuki, K., et al. 2003. Biochem. Biophys. Res. Commun.309, 768.
Kawabata, K., et al. 1991. Biochem. Biophys. Res. Commun.177, 814.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
324759-1MG04055977216080