Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
UNSPSC Code:
41116012
NACRES:
NB.22
eCl@ss:
32160405
Pricing and availability is not currently available.
form
liquid
Quality Segment
manufacturer/tradename
Chemicon®
technique(s)
DNA amplification: suitable
shipped in
ambient
General description
Nuclease Free water. Liquid is RNase and DNase Free.
Nuclease-free Water is essential to preserve the sample integrity for applications such as PCR, cloning, sequencing, or gene editing. DEPC chemical treatment is often used to eliminate the nucleases from solutions as it is an efficient inhibitor of DNase and RNase.
Nuclease-free Water is essential to preserve the sample integrity for applications such as PCR, cloning, sequencing, or gene editing. DEPC chemical treatment is often used to eliminate the nucleases from solutions as it is an efficient inhibitor of DNase and RNase.
Application
Suitable for use in polymerase chain reaction and other molecular biology techniques.
Packaging
5x2 mL
Preparation Note
Store at room temp
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
1 of 1
This Item | |||
|---|---|---|---|
| description - | description OmniPur® Grade, DEPC Treated, Autoclaved, Nuclease-Free | description OmniPur® Grade, Sterile, Nuclease Free | description Microbial DNA-free; Suitable for PCR, sequencing and NGS |
| technique(s) DNA amplification: suitable | technique(s) - | technique(s) - | technique(s) DNA extraction: suitable, DNA sequencing: suitable, PCR: suitable |
| Quality Level 300 | Quality Level 100 | Quality Level 100 | Quality Level 200 |
| form liquid | form liquid | form liquid | form liquid |
| shipped in ambient | shipped in ambient | shipped in ambient | shipped in ambient |
| manufacturer/tradename Chemicon® | manufacturer/tradename Calbiochem® | manufacturer/tradename Calbiochem® | manufacturer/tradename - |
Storage Class
10 - Combustible liquids
wgk
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


