Skip to Content
Merck

3096

Bacterial Xpress Nucleic Acid Extraction Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

eCl@ss:
32161000
UNSPSC Code:
12161503
NACRES:
NA.51
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


manufacturer/tradename

Chemicon®

Quality Segment

technique(s)

DNA extraction: suitable

application(s)

sample preparation

shipped in

wet ice

General description

Intended Use

The BacterialXpress nucleic acid extraction kit is intended for the purification of bacterial DNA from plasma, serum, cerebral spinal fluid, amniotic fluid, tissue, bone marrow, and cell culture. The BacterialXpress kit provides a rapid method for the extraction and purification of amplification-competent nucleic acids; however, the kit does not separate DNA from RNA. The BacterialXpress nucleic acid extraction kit can be used for the extraction and purification of a broad range of bacterial DNA from a variety of sources. However, performance cannot be guaranteed for all bacteria from all sources.

FOR LABORATORY USE ONLY; not for use in diagnostic procedures.

Application

Research Category
All
Suitable for the purification of bacterial DNA from plasma, serum, cerebral spinal fluid, amniotic fluid, tissue, bone marrow, and cell culture.

Packaging

50 Extractions

Preparation Note

BacterialXpress references a 2-8°C storage temperature, however, the product can also be stored at room temperature upon arrival. Product is stable up to the expiration date printed on the kit label at 2-8°C or at room temperature. Reagent will crystallize upon storage at 2-8°C and should be re-dissolved with gentle warming at 37° C before use.

WARNINGS AND PRECAUTIONS:

The BacterialXpress nucleic acid extraction reagent removes endogenous nucleases. Certified nuclease-free plasticware should be used whenever possible and work surfaces should be cleaned frequently with a diluted bleach solution to reduce the possibility for nuclease contamination.

Proper aseptic technique should be used when working with potentially infectious material. The BacterialXpress reagent contains a chaotropic salt, which is an irritant and is not compatible with disinfecting agents that contain bleach.

Dispose chaotropic salt containing solutions and plasticware according to your institutions waste-disposal guidelines.

The BacterialXpress reagent contains chaotropic salts, which may irritate the skin and damage clothing. As with any extraction procedure, use gloves, a laboratory coat and safety goggles. Avoid breathing vapors. In case of contact, immediately flush eyes or skin with a large amount of saline or water for at least 15 minutes.

DO NOT ALLOW THE EXTRACTION REAGENT TO COME IN CONTACT WITH BLEACH.

Other Notes

Bacterial DNA Extraction reagent - (Part No. 5539) - (1 bottle, 11.0 mL).

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

General Lab Use


pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1C

supp_hazards

Storage Class

8A - Combustible corrosive hazardous materials

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library