Skip to Content
Merck
All Photos(1)

Key Documents

287810

Sigma-Aldrich

2′,7′-Dichlorofluorescin Diacetate

Cell-permeable fluorogenic probe that is useful for the detection of reactive oxygen species (ROS) and nitric oxide (•NO) and for the determination of the degree of overall oxidative stress.

Synonym(s):

2′,7′-Dichlorofluorescin Diacetate, H₂DCFDA, 2ʹ,7ʹ-Dichlorodihydrofluorescein Diacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H16Cl2O7
CAS Number:
Molecular Weight:
487.29
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.25

Quality Level

Assay

≥94% (HPLC)

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

Clc1cc2c(cc1OC(=O)C)Oc3c(cc(c(c3)OC(=O)C)Cl)C2c4c(cccc4)C(=O)O

InChI

1S/C24H16Cl2O7/c1-11(27)31-21-9-19-15(7-17(21)25)23(13-5-3-4-6-14(13)24(29)30)16-8-18(26)22(32-12(2)28)10-20(16)33-19/h3-10,23H,1-2H3,(H,29,30)

InChI key

PXEZTIWVRVSYOK-UHFFFAOYSA-N

General description

Cell-permeable fluorogenic probe that is useful for the detection of reactive oxygen species (ROS) and the nitric oxide (NO) in neutrophils and macrophages and for the determination of the overall oxidative stress in toxicological phenomena.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Detection of reactive oxygen species (ROS) and nitric oxide (•NO)
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Crow, J.P. 1997. Nitric Oxide 1, 145.
Ripple, M.O., et al. 1997. J. Natl. Cancer Inst. 89, 40.
Caro, A., and Puntarulo, S. 1996. Biochim. Biophys. Acta1291, 245.
Perez-Garcia, F., et al. 1996. Life Sci. 59, 2033.
Ramakrishnan, N., et al. 1996. Biochem. Pharmacol. 51, 1443.
Ferrer, A.S., et al. 1990. Anal. Biochem. 187, 129.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hong-Wei Jiang et al.
Frontiers in physiology, 12, 662666-662666 (2021-07-16)
The aim of this study was to explore the protective effects and the regulatory mechanisms of bariatric surgery on kidney injury in diabetic rats. We established a useful type 2 diabetic rat model using high-fat and high-sugar diet feeding following
Thivanka Muthumalage et al.
Toxics, 8(3) (2020-07-02)
Recently, there has been an outbreak associated with the use of e-cigarette or vaping products, associated lung injury (EVALI). The primary components of vaping products, vitamin E acetate (VEA) and medium-chain triglycerides (MCT), may be responsible for acute lung toxicity.
Shaiesh Yogeswaran et al.
Toxics, 9(10) (2021-10-23)
Studies have shown that aerosols generated from flavored e-cigarettes contain Reactive Oxygen Species (ROS), promoting oxidative stress-induced damage within pulmonary cells. Our lab investigated the ROS content of e-cigarette vapor generated from disposable flavored e-cigarettes (vape bars) with and without
Noviana Wulansari et al.
Science advances, 7(8) (2021-02-19)
Loss-of-function mutations of DNAJC6, encoding HSP40 auxilin, have recently been identified in patients with early-onset Parkinson's disease (PD). To study the roles of DNAJC6 in PD pathogenesis, we used human embryonic stem cells with CRISPR-Cas9-mediated gene editing. Here, we show
Hui-Hui Wang et al.
Frontiers in endocrinology, 12, 670031-670031 (2021-07-23)
Ferroptosis is an emerging programmed cell death distinguished from apoptosis and autophagy and plays essential roles in tumorigenesis. Thyroid cancer is a prevalent endocrine tumor, but the molecular mechanism of ferroptosis during thyroid cancer development remains unclear. Here, we identified

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service