Skip to Content
Merck

233490

Coagulation Factor XII, Human Plasma

Coagulation Factor XII, Human Plasma, CAS 9001-30-3, is a native coagulation factor XI that once activated (principally by kallikrein), is converted to Factor α-XIIa.

Synonym(s):

Coagulation Factor XII, Human Plasma

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
MDL number:
NACRES:
NA.42
UNSPSC Code:
12352202
Form:
liquid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

description

Merck USA index - 14, 3925

form

liquid

specific activity

≥20 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−70°C

General description

Coagulation Factor XII, Human Plasma, CAS 9001-30-3, is a native coagulation factor XI that once activated (principally by kallikrein), is converted to Factor α-XIIa.
Native coagulation factor XII from human plasma. A single-chain polypeptide, which once activated (principally from the action of kallikrein) is converted to Factor α-XIIa. Factor α-XIIa then converts Factor XI to Factor XIa.

Packaging

Please refer to vial label for lot-specific concentration.

Physical form

In 150 mM NaCl, 4 mM sodium acetate buffer, pH 5.3.

Preparation Note

Following initial thaw, aliquot and freeze (-70°C).
Prepared from plasma that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Analysis Note

single band by SDS-PAGE

Other Notes

Specific activity is measured by clotting assay and compared to a standard curve produced using normal pooled human plasma.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library