Skip to Content
Merck

229103

Choline Kinase-a Inhibitor, CK37

The Choline Kinase-α Inhibitor, CK37, also referenced under CAS 1001478-90-5, controls the biological activity of Choline Kinase-α. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Choline Kinase-a Inhibitor, CK37, CK37, N-(3,5-dimethyl-dimethylphenyl)-2-[[5-(4-ethylphenyl)-1H-1,2,4-triazol-3-yl]sulfanyl]acetamide, N-(3,5-dimethyl-dimethylphenyl)-2-[[5-(4-ethylphenyl)-1H-1,2,4-triazol-3-yl]sulfanyl]acetamide, CK37

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H22N4OS
CAS Number:
Molecular Weight:
366.48
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥95% (HPLC)
Form:
powder
Storage condition:
OK to freeze, protect from light
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

tan

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

O=C(NC1=CC(C)=CC(C)=C1)CSC2=NNC(C3=CC=C(CC)C=C3)=N2

InChI

1S/C20H22N4OS/c1-4-15-5-7-16(8-6-15)19-22-20(24-23-19)26-12-18(25)21-17-10-13(2)9-14(3)11-17/h5-11H,4,12H2,1-3H3,(H,21,25)(H,22,23,24)

InChI key

PSJVIOBOAOSNIY-UHFFFAOYSA-N

General description

A cell-permeable, non-Hemicholinium-3 derivative and choline-competitive choline kinase-α inhibitor. It is demonstrated to inhibit endogenous choline kinase activity, dose-dependently, between 1-10 µM, and dose-dependently decrease phosphocholine concentrations, as well as levels of downstream choline metabolites, including phosphatidylcholine and phosphatidic acid in HeLa cells. At 10 µM, it is also shown to attenuate MAPK and PI3K/AKT signaling, and disrupt the actin cytoskeleton and membrane ruffling, consistent with siRNA choline kinase-&alpha transfection. This compound exhibits dose-dependent suppression of cell growth in six tumor cell lines (IC50 = 5-10 µM), in vitro. Furthermore, it is shown to suppress tumor growth in a mouse model by 48% at 0.08 mg/g daily dose for 8 days without obvious toxicity, in vivo.

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°c.

Other Notes

Clem, B., et al. 2011. Oncogene30, 3370.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library