Skip to Content
Merck
All Photos(1)

Key Documents

204881

Sigma-Aldrich

Complement C3a Peptide

≥95% (SDS-PAGE), liquid, human

Synonym(s):

C3a Protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

Product Name

Complement C3a, Human, Complement C3a, Human, is a native, single chain C3a complement component. It has many biological activities, such as smooth muscle contraction, platelet activation, & immunoregulatory reactions.

Quality Level

Assay

≥95% (SDS-PAGE)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

impurities

≤10% C3adesArg

shipped in

wet ice

storage temp.

−70°C

General description

Native, human C3a complement component. Single chain, 77 amino acid peptide. Activation of either complement pathway can result in formation of C3-cleaving enzyme (C3 convertases) on target surfaces. The C3 convertase enzymes of either complement activation pathway will cleave the C3 α-chain at peptide bond 77, resulting in the production and release of the C3a peptide. C3a, one of the three complement-derived anaphylatoxins, expresses a wide variety of biological activities, which include smooth muscle contraction, platelet and neutrophil activation and aggregation, skin wheal and flare, and immunoregulatory reactions.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In PBS, pH 7.2, sterile-filtered through a 0.22 µm filter.

Preparation Note

Prepared from serum that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Hugli, T.E. 1989. Curr. Top. Micro. Immunol. 153, 181.
Hugli, T.E. 1986. Complement 3, 111.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service