Skip to Content
Merck

201102

Bilirubin Conjugate, Ditaurate, Disodium Salt

solid, bilirubin standard, Calbiochem®

Synonym(s):

Bilirubin ditaurate (BDT)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352200
NACRES:
NA.77
Form:
solid
Quality level:
Storage condition:
OK to freeze, desiccated (hygroscopic), protect from light
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Bilirubin Conjugate, Ditaurate, Disodium Salt, Bilirubin Conjugate, Ditaurate, Disodium Salt is suitable as a direct bilirubin standard. Behaves similar to purified bilirubin glucuronides from human bile in diazo tests. Unstable in solution.

Quality Level

description

RTECS - DU3038000

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, desiccated (hygroscopic), protect from light

solubility

water: 10 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

Bilirubin ditaurate (BDT) is the first bile pigment found naturally in the biles of specific fish species. It consists of unconjugated bilirubin (UCB) with taurine side chains attached to the C8 and C12 propionic acids, similar in length to glucuronic acids.

Application

Bilirubin Conjugate, Ditaurate, Disodium Salt has been used to spike patient samples to access icterus interference. It has also been used to prepare test pools for conjugated bilirubin to screen bilirubin interference.
Bilirubin Conjugate, Ditaurate, Disodium Salt – Calbiochem has been used as a standard for testing the interference of conjugated bilirubin in assays aimed at quantifying analytes in serum or plasma samples.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Suitable as a direct bilirubin standard
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Preparation Note

Unstable in solution; reconstitute just prior to use.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



A novel canine nuclear magnetic resonance spectroscopy-based metabolomics platform: Validation and sample handling
Ottka C, et al.
Veterinary Clinical Pathology / American Society For Veterinary Clinical Pathology (2021)
Self-assembly of aqueous bilirubin ditaurate, a natural conjugated bile pigment, to contraposing enantiomeric dimers and M(-) and P(+) tetramers and their selective hydrophilic disaggregation by monomers and micelles of bile salts
Neubrand MW, et al.
Biochemistry (2015)
Rising above helium: A hydrogen carrier gas chromatography flame ionization detection (GC-FID) method for the simultaneous quantification of toxic alcohols and ethylene glycol in human plasma specimens
Buse J, et al.
Clinical Biochemistry (2019)



Global Trade Item Number

SKUGTIN
201102-100MG04055977221589