Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
41116012
NACRES:
NA.41
eCl@ss:
32160405
form
liquid
Quality Segment
manufacturer/tradename
Upstate®
technique(s)
activity assay: suitable (phosphatase)
shipped in
wet ice
Application
For use in the malachite green assay kit.
Research Category
All
All
Physical form
0.5ml of 1% Tween®-20. Liquid at 4°C.
Preparation Note
2 years at 4°C
Analysis Note
Routinely evaluated by Malachite Green Assay.
Legal Information
TWEEN is a registered trademark of Croda International PLC
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class
10 - Combustible liquids
wgk
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
ELL-associated factor 2 (EAF2), a functional homolog of EAF1 with alternative ELL binding properties.
Federico Simone,Roger T Luo,Paul E Polak,Joseph J Kaberlein,Michael J Thirman
Blood null
M H Chen et al.
The Journal of biological chemistry, 270(44), 26168-26177 (1995-11-03)
alpha-Agglutinin of Saccharomyces cerevisiae is a cell wall-associated protein that mediates cell interaction in mating. Although the mature protein includes about 610 residues, the NH2-terminal half of the protein is sufficient for binding to its ligand a-agglutinin. alpha-Agglutinin20-351, a fully
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 20-104 | 04053252371295 |