Skip to Content

178680

L(+)Arabinose

A pectin sugar widely distributed in plants, usually in the form of complex polysaccharides.

Synonym(s):

L(+)Arabinose

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C5H10O5
CAS Number:
Molecular Weight:
150.13
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.79
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

description

Merck USA index - 14, 761

assay

≥99% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: 100 mg/mL

shipped in

ambient

storage temp.

15-25°C

SMILES string

O1C([C@H]([C@@H]([C@@H](C1)O)O)O)O

InChI

1S/C5H10O5/c6-2-1-10-5(9)4(8)3(2)7/h2-9H,1H2/t2-,3-,4+,5?/m1/s1

InChI key

SRBFZHDQGSBBOR-ZRMNMSDTSA-N

General description

A pectin sugar widely distributed in plants, usually in the form of complex polysaccharides.

Preparation Note

Following reconstitution, filter-sterilize and store at room temperature (20°C). Stock solutions are stable for up to 6 months at room temperature.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library