Skip to Content
Merck
All Photos(1)

Key Documents

178467

Sigma-Aldrich

Anti-Apolipoprotein B Goat pAb

liquid, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

goat

Quality Level

antibody form

diluted serum

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Anti-Apolipoprotein B, goat polyclonal, recognizes human apolipoprotein B. It is validated for Western blotting, immunoelectrophoresis, and immunoprecipitation.
Goat polyclonal antibody supplied as diluted serum that has been defibrinated, delipidized, and adsorbed by solid phase chromatography. Recognizes apolipoprotein B.
Recognizes apolipoprotein B.
  • Antibody Target Gene Symbol: APOB
  • Target Synonym: Aa1064, Ac1-060, AI315052, ApoB-100, APOB-29, ApoB-48, APOLIPOPROTEIN B, APOLIPOPROTEIN B100, FLDB, LDLCQ4, MGC176318
  • Entrez Gene Name: apolipoprotein B (including Ag(x) antigen)
  • Hu Entrez ID: 338 (Related Antibodies: 178446, 178444)
  • Mu Entrez ID: 238055
  • Rat Entrez ID: 54225
  • Immunogen

    Human
    purified human, apolipoprotein B

    Application

    Immunoblotting (see application references)

    Immunoelectrophoresis (see comments)

    Immunoprecipitation (Becker titer, ≥21 mg/ml)

    Packaging

    Please refer to vial label for lot-specific concentration.

    Warning

    Toxicity: Standard Handling (A)

    Physical form

    Serum diluted in 500 mM NaCl, 50 mM Tris-HCl, pH 7.5.

    Reconstitution

    Following initial thaw, aliquot and freeze (-20°C).

    Other Notes

    Monospecific as determined by immunoelectrophoresis (IEP) against 2X pooled human serum and neat pooled human plasma. Variables associated with assay conditions will dictate the proper working dilution.

    Legal Information

    CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

    Not finding the right product?  

    Try our Product Selector Tool.

    Storage Class Code

    10 - Combustible liquids

    WGK

    WGK 1

    Flash Point(F)

    Not applicable

    Flash Point(C)

    Not applicable


    Certificates of Analysis (COA)

    Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Tanja Kosenko et al.
    The Journal of biological chemistry, 288(12), 8279-8288 (2013-02-13)
    Proprotein convertase subtilisin/kexin type-9 (PCSK9) is a secreted protein that binds to the epidermal growth factor-like-A domain of the low density lipoprotein receptor (LDLR) and mediates LDLR degradation in liver. Gain-of-function mutations in PCSK9 are associated with autosomal dominant hypercholesterolemia
    Yuehui Wang et al.
    PloS one, 4(12), e8442-e8442 (2009-12-31)
    A small fraction of dietary protein survives enzymatic degradation and is absorbed in potentially antigenic form. This can trigger inflammatory responses in patients with celiac disease or food allergies, but typically induces systemic immunological tolerance (oral tolerance). At present it
    David Melville et al.
    Molecular biology of the cell, 30(3), 387-399 (2018-11-30)
    COPII-coated vesicles are the primary mediators of ER-to-Golgi trafficking. Sar1, one of the five core COPII components, is a highly conserved small GTPase, which, upon GTP binding, recruits the other COPII proteins to the ER membrane. It has been hypothesized
    David B Melville et al.
    The Journal of biological chemistry, 295(25), 8401-8412 (2020-05-03)
    Vesicles that are coated by coat protein complex II (COPII) are the primary mediators of vesicular traffic from the endoplasmic reticulum to the Golgi apparatus. Secretion-associated Ras-related GTPase 1 (SAR1) is a small GTPase that is part of COPII and
    Enrico D Perini et al.
    Traffic (Copenhagen, Denmark), 15(12), 1366-1389 (2014-10-01)
    Early endosomes are organized in a network of vesicles shaped by cycles of fusion, fission, and conversion to late endosomes. In yeast, endosome fusion and conversion are regulated, among others, by CORVET, a hexameric protein complex. In the mammalian endocytic

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service