Skip to Content
Merck
All Photos(10)

Key Documents

1.73027

Supelco

Prove 300 plus

photometric, 190-1100 nm, Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
SE.11
Pricing and availability is not currently available.

product line

Spectroquant®

Quality Level

packaging

pkg of 1 unit

measuring range

190-1100 nm

technique(s)

UV/Vis spectroscopy: suitable
photometry: suitable

compatibility

for use with Spectroquant® (Test kits)
for use with Spectroquant®

detection method

photometric (UV-Vis spectroscopy)

storage temp.

10-30°C

General description

The Spectroquant Prove 300 plus is the best choice for those who primarily use our broad range of Spectroquant Test Kits, or only perform UV/Vis measurements. The Spectroquant Prove 300 plus provides high quality and great value for money for your daily analyses. The Spectroquant Prove plus family is our series of cutting-edge spectrophotometers for easy analysis of water, food and beverage, and environmental samples. All Spectroquant Prove plus instruments are pre-programmed for over 200 Test Kits and automatic bar code recognition ensures the correct method parameters are applied and all essential information such as lot number, expiry date, and automatic calibration updates are recorded, making the use of analytical Test Kits as easy as never before. UV/VIS Spectrophotometer 4 nm spectral bandwidth Spectroquant®. The Spectroquant® Prove plus photometers are upgraded highlighting smoother functionality with instant responsiveness, a more receptive display with faster processing, and increased storage capacity.

Application

Spectroquant® Prove plus photometers find application in



  • Wastewater testing
  • Drinking water testing
  • Process water testing
  • Environmental testing (groundwater, soil analysis)
  • Food & Beverage testing (brewery methods, sugar colors, oil determinations, bottled water, etc.)

Features and Benefits

Pre-programmed to work seamlessly with more than 150 Spectroquant® test kits, Spectroquant® Prove plus photometers are among the most trusted for your analysis.



  • Enhanced functionality for seamless operation and instant responsiveness
  • Improved display responsiveness and faster processing capabilities
  • Expanded storage capacity, allowing for storage of up to 7000 standard measurement results
  • Extended warranty period of 2 years
  • Easy to use and pre-programmed with pre-defined reagents
  • Reduced use of chemicals compared to traditional methods
  • Suitable for a broad measuring range per parameter (minimizes dilution work)
  • Pre-programmed for over 300 analysis methods
  • Instrument-supported AQA system for easy quality assurance
  • Automatic cell size recognition for cell & reagent tests
  • Live ID barcode for all cell & reagent tests (incl. lot and expiry data)
  • No adapter is needed for the measurement of different cell sizes
  • Kits can be conveniently stored at room temperature

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Discover the K270 method for analyzing olive oil quality as per EEC Regulation 2568/91. Access detailed protocols and ensure compliance with standards.

Discover the K270 method for analyzing olive oil quality as per EEC Regulation 2568/91. Access detailed protocols and ensure compliance with standards.

Discover the K270 method for analyzing olive oil quality as per EEC Regulation 2568/91. Access detailed protocols and ensure compliance with standards.

Discover the K270 method for analyzing olive oil quality as per EEC Regulation 2568/91. Access detailed protocols and ensure compliance with standards.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service