Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.24
UNSPSC Code:
41116107
grade
certified reference material
Quality Segment
product line
Certipur®
concentration
1000 mg/L Sm in 2-3% HNO3
technique(s)
ICP: suitable
pH
0.5 (20 °C in H2O)
density
1.013 g/cm3 at 20 °C
application(s)
industrial qc
pharmaceutical
format
single component solution
storage temp.
15-25°C
General description
This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from a National Metrology Institute (NMI), e.g. NIST.
Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.
Download your certificate at https://www.sigma-aldrich.com to view certified values, including uncertainty, date of expiry, and detailed information about trace impurities.
Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.
Download your certificate at https://www.sigma-aldrich.com to view certified values, including uncertainty, date of expiry, and detailed information about trace impurities.
Application
Samarium standard can beused as a calibration standard in the determination of samarium inenvironmental samples by inductively coupled plasma optical emissionspectroscopy (ICP-OES)
Other Notes
For a complete product listing of our ICP and AAS range, including technical information and example certificates, please visit ICP & AAS standards website.
Detailed information on our complete workflow solutions for ICP-AAS can be found in our Brochure: High-Purity Chemicals for Inorganic Trace Analysis.
In our effort to be more sustainable, we′re going digital with COAs-download them directly from our website.
Detailed information on our complete workflow solutions for ICP-AAS can be found in our Brochure: High-Purity Chemicals for Inorganic Trace Analysis.
In our effort to be more sustainable, we′re going digital with COAs-download them directly from our website.
Legal Information
CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2
wgk
nwg
Storage Class
12 - Non Combustible Liquids
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
