Skip to Content
Merck
All Photos(1)

Key Documents

164610

Sigma-Aldrich

p-Aminophenylmercuric Acetate

An organomercurial agent used for the activation of latent MMPs in vitro. Acts by facilitating the loss of the enzyme propeptide domain through an autolytic cleavage known as the cysteine switch.

Synonym(s):

p-Aminophenylmercuric Acetate, APMA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9HgNO2
CAS Number:
Molecular Weight:
351.75
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

description

RTECS - OV5550000

Assay

≥90% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

yellow

solubility

0.1 M NaOH: 10 mg/mL
DMSO: 10 mg/mL

shipped in

ambient

storage temp.

10-30°C

SMILES string

[Hg+]c1ccc(cc1)N.[O-]C(=O)C

InChI

1S/C6H6N.C2H4O2.Hg/c7-6-4-2-1-3-5-6;1-2(3)4;/h2-5H,7H2;1H3,(H,3,4);/q;;+1/p-1

InChI key

RXSUFCOOZSGWSW-UHFFFAOYSA-M

General description

An organomercurial agent used for the activation of latent MMPs in vitro. Acts by facilitating the loss of the enzyme propeptide domain through an autolytic cleavage known as the cysteine switch.
An organomercurial agent used for the activation of latent matrix metalloproteinases (MMPs) in vitro. Acts by facilitating the loss of the enzyme propeptide domain through an autolytic cleavage known as the cysteine switch.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Activation of latent MMPs in vitro
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Highly Toxic (H)

Reconstitution

Following reconstitution, refrigerate (4°C). Stock solutions are stable for up to 1 month at 4°C.

Other Notes

Chen, J.-M., et al. 1991. J. Biol. Chem. 266, 5113.
Marcy, A.I., et al. 1991. Biochemistry30, 6476.
Stricklin, G.P., et al. 1983. Biochemistry22, 61.
Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Saskia V Vijver et al.
Frontiers in immunology, 12, 733561-733561 (2021-10-26)
The tumor microenvironment (TME) is a complex structure comprised of tumor, immune and stromal cells, vasculature, and extracellular matrix (ECM). During tumor development, ECM homeostasis is dysregulated. Collagen remodeling by matrix metalloproteinases (MMPs) generates specific collagen fragments, that can be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service